Få mere at vide
Methoxyfenozide, TRC
Artikelnummer. 30004973
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004973 10 g
17932825 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004973 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26276510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 161050-58-4
Molekylær formel C22 H28 N2 O3
Molekylvægt (g/mol) 368.47
Synonym Methoxyfenozide, Benzoic acid, 3-methoxy-2-methyl-, 2-(3,5-dimethylbenzoyl)-2-(1,1-dimethylethyl)hydrazide (9CI, ACI), 3,5-Dimethylbenzoic acid N-tert-butyl-N-(3-methoxy-2-methylbenzoyl)hydrazide, Intrepid, Intrepid (pesticide), Methoxyfenozide, Methoxyphenozide, Prodigy 240SC, RG 102398, RH 112485, RH 2485, Runner
IUPAC navn N'-tert-butyl-N'-(3,5-dimethylbenzoyl)-3-methoxy-2-methylbenzohydrazide
SMIL COc1cccc(C(=O)NN(C(=O)c2cc(C)cc(C)c2)C(C)(C)C)c1C
Tekniske data
Kemisk navn eller materiale Methoxyfenozide
Renhedsgrad noter HPLC
Mængde 10 g
InChI formel InChI=1S/C22H28N2O3/c1-14-11-15(2)13-17(12-14)21(26)24(22(4,5)6)23-20(25)18-9-8-10-19(27-7)16(18)3/h8-13H,1-7H3,(H,23,25)
Formel vægt 368.21
Procent renhed >95%