Learn More
Methoxyacetonitrile, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 1738-36-9 |
| Molecular Formula | C3H5NO |
| MDL Number | MFCD00001892 |
| Quantity | 1 g |
| SMILES | N#CCOC |
| Molecular Weight (g/mol) | 71.08 |
| Percent Purity | 97% |
| Chemical Name or Material | Methoxyacetonitrile |