Få mere at vide
Methoxy(monomethyl Carbonato-O')-magnesium (~0.4M in Methanol), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methoxy(monomethyl Carbonato-O')-magnesium |
| CAS | 4861-79-4 |
| Molekylær formel | C3H6MgO4 |
| Mængde | 100 mL |
| Synonym | Magnesium Methyl Carbonate, (Hydrogen carbonato)methoxy-magnesium methyl ester, Carbonic acid monomethyl ester magnesium complex, Methoxymagnesium methyl carbonate, Methyl methoxymagnesium carbonate, MMC, Stile’s reagent |
| InChI formel | InChI=1S/C2H4O3.CH3O.Mg/c1-5-2(3)4;1-2;/h1H3,(H,3,4);1H3;/q;-1;+2/p-1 |
| SMIL | [O-]C(OC)=O.[O-]C.[Mg+2] |
| IUPAC navn | magnesium methanolate methyl carbonate |
| Molekylvægt (g/mol) | 130.38 |
| Formel vægt | 130.01 |
| Vis mere |