Få mere at vide
Methoxy Eltrombopag, TRC
Artikelnummer. 30132775
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132775 250 mg
30138297 100 mg
30012231 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132775 Leverandør Toronto Research Chemicals Leverandørnr. TRCM493100250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1437383-35-1
Molekylær formel C26 H24 N4 O4
Molekylvægt (g/mol) 456.49
Synonym 3'-[(2Z)-2-[1-(3,4-Dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-methoxy-[1,1'-biphenyl]-3-carboxylic acid
IUPAC navn 3-[3-[(2Z)-2-[1-(3,4-dimethylphenyl)-3-methyl-5-oxo-pyrazol-4-ylidene]hydrazino]-2-methoxy-phenyl]benzoic acid
SMIL COc1c(N\N=C/2\C(=NN(C2=O)c3ccc(C)c(C)c3)C)cccc1c4cccc(c4)C(=O)O
Tekniske data
Kemisk navn eller materiale Methoxy Eltrombopag
Mængde 250 mg
InChI formel InChI=1S/C26H24N4O4/c1-15-11-12-20(13-16(15)2)30-25(31)23(17(3)29-30)28-27-22-10-6-9-21(24(22)34-4)18-7-5-8-19(14-18)26(32)33/h5-14,27H,1-4H3,(H,32,33)/b28-23-
Formel vægt 456.1798