Få mere at vide
Methoxamine Hydrochloride, LoGiCal
Artikelnummer. 30459288
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459288 10 mg
1 options
Artikelnummer. 30459288 Leverandør LoGiCal Leverandørnr. LGCFOR1062.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methoxamine Hydrochloride
CAS 61-16-5
Analyt- eller komponentnavne Methoxamine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C11 H17 N O3 . Cl H
Certificeringer/compliance ISO 9001
Synonym Methoxamine hydrochloride, 2-Amino-1-(2,5-dimethoxyphenyl)-1-propanol hydrochloride, 2-Amino-1-hydroxy-1-(2,5-dimethoxyphenyl)propane hydrochloride, Beta-hydroxy-beta-(2,5-dimethoxyphenol)-isopropylamine hydrochloride, Pressomin hydrochloride, Vasoxine, Vasoxine hydrochloride, Vasoxyl, Vasoxyl hydrochloride, Vasylox, Benzenemethanol, alpha-(1-aminoethyl)-2,5-dimethoxy-, hydrochloride, Benzyl alcohol, alpha-(1-aminoethyl)-2,5-dimethoxy-, hydrochloride, alpha-(alpha-Aminoethyl)-2,5-dimethoxybenzyl alcohol hydrochloride, beta-Hydroxy-beta-(2,5-dimethoxyphenyl)isopropylamine hydrochloride
InChI formel InChI=1S/C11H17NO3.ClH/c1-7(12)11(13)9-6-8(14-2)4-5-10(9)15-3;/h4-7,11,13H,12H2,1-3H3;1H
Løsningstype Neat
SMIL Cl.COc1ccc(OC)c(c1)C(O)C(C)N
IUPAC navn 2-amino-1-(2,5-dimethoxyphenyl)propan-1-ol;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 247.72
Mængde 10 mg
Formel vægt 247.0975 g/mol