Få mere at vide
Methotrexate Dimethyl Ester, TRC
Artikelnummer. 30136121
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136121 500 mg
30137748 250 mg
17963645 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136121 Leverandør Toronto Research Chemicals Leverandørnr. TRCM260685500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34378-65-9
Molekylær formel C22 H26 N8 O5
Molekylvægt (g/mol) 482.49
Synonym L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, 1,5-dimethyl ester, Glutamic acid, N-[p-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, dimethyl ester, L-(+)- (8CI), L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, dimethyl ester (9CI), Dimethyl methotrexate, Methotrexate dimethyl ester, Methotrexate Imp. J (EP)
IUPAC navn dimethyl (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioate
SMIL COC(=O)CC[C@H](NC(=O)c1ccc(cc1)N(C)Cc2cnc3nc(N)nc(N)c3n2)C(=O)OC
Tekniske data
Kemisk navn eller materiale Methotrexate Dimethyl Ester
Renhedsgrad noter HPLC
InChI formel InChI=1S/C22H26N8O5/c1-30(11-13-10-25-19-17(26-13)18(23)28-22(24)29-19)14-6-4-12(5-7-14)20(32)27-15(21(33)35-3)8-9-16(31)34-2/h4-7,10,15H,8-9,11H2,1-3H3,(H,27,32)(H4,23,24,25,28,29)/t15-/m0/s1
Mængde 500 mg
Formel vægt 482.2026
Procent renhed >95%