Få mere at vide
Methotrexate-d3 Dimethyl Ester, TRC
Artikelnummer. 30467907
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30467907 2.5 mg 2.5mg
30465360 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467907 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2606872.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 432545-60-3
Molekylær formel C22 2H3 H23 N8 O5
Molekylvægt (g/mol) 485.51
Synonym L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methyl-d3-amino]benzoyl]-, dimethyl ester (9CI)
IUPAC navn dimethyl (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-(trideuteriomethyl)amino]benzoyl]amino]pentanedioate
SMIL [2H]C([2H])([2H])N(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)OC)C(=O)OC
Tekniske data
Kemisk navn eller materiale Methotrexate-methyl-d3, Dimethyl Ester
Mængde 2.5 mg
InChI formel InChI=1S/C22H26N8O5/c1-30(11-13-10-25-19-17(26-13)18(23)28-22(24)29-19)14-6-4-12(5-7-14)20(32)27-15(21(33)35-3)8-9-16(31)34-2/h4-7,10,15H,8-9,11H2,1-3H3,(H,27,32)(H4,23,24,25,28,29)/t15-/m0/s1/i1D3
Formel vægt 485.2214