Få mere at vide
Methoprene, TRC
Artikelnummer. 30180314
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180314 100 mg
30180350 250 mg
30179848 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180314 Leverandør Toronto Research Chemicals Leverandørnr. TRCM260715100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 40596-69-8
Molekylær formel C19 H34 O3
Molekylvægt (g/mol) 310.47
Synonym 2,4-Dodecadienoic acid, 11-methoxy-3,7,11-trimethyl-, 1-methylethyl ester, (E,E)-, (2E,4E)-3,7,11-Trimethyl-11-methoxydodeca-2,4-dienoic acid ethyl ester, Altosid, Altosid Liquid Larvicide, Altosid SR 10, Altosid XR, Apex, Apex (pesticide), Apex 5E, Apex 600E, Bioprene BM Fire Ant Killer Bait, Dianex, ENT 70460, Isopropyl (2E,4E)-11-methoxy-3,7,11-trimethyl-2,4-dodecadienoate, Juvemon, Juvenmon, Kabat, Manina, Manta, Manta (hormone), Methoprene, Methoprene S, Metoprag 20CE, SB-515, Starbar Inhibitor, ZPA 1019, ZR 515, dl-Isopropyl 11-methoxy-3,7,11-trimethyl-trans-trans-2,4-dodecadienoate
IUPAC navn propan-2-yl (2E,4E)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
SMIL COC(C)(C)CCCC(C)C\C=C\C(=C\C(=O)OC(C)C)\C
Tekniske data
Kemisk navn eller materiale Methoprene
Mængde 100 mg
InChI formel InChI=1S/C19H34O3/c1-15(2)22-18(20)14-17(4)11-8-10-16(3)12-9-13-19(5,6)21-7/h8,11,14-16H,9-10,12-13H2,1-7H3/b11-8+,17-14+
Formel vægt 310.2508