Få mere at vide
Methohexital (Mixture of Diastereomers), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methohexital |
| CAS | 151-83-7 |
| Molekylær formel | C14 H18 N2 O3 |
| Mængde | 100 mg |
| Synonym | 2,4,6(1H,3H,5H)-Pyrimidinetrione, 1-methyl-5-(1-methyl-2-pentyn-1-yl)-5-(2-propen-1-yl)- (ACI), 1-Methyl-5-(1-methyl-2-pentyn-1-yl)-5-(2-propen-1-yl)-2,4,6(1H,3H,5H)-pyrimidinetrione (ACI), 2,4,6(1H,3H,5H)-Pyrimidinetrione, 1-methyl-5-(1-methyl-2-pentynyl)-5-(2-propenyl)- (9CI), Barbituric acid, 5-allyl-1-methyl-5-(1-methyl-2-pentynyl)- (6CI, 7CI, 8CI), 5-Allyl-5-(3-hexyn-2-yl)-1-methylbarbituric acid, Brevital, Brietal, Compound 22451, Compound 25398, Enallynymall, MeSH ID: D008723, Methodrexitone, Methohexital, Methohexitone |
| InChI formel | InChI=1S/C14H18N2O3/c1-5-7-8-10(3)14(9-6-2)11(17)15-13(19)16(4)12(14)18/h6,10H,2,5,9H2,1,3-4H3,(H,15,17,19) |
| SMIL | CCC#CC(C)C1(CC=C)C(=O)NC(=O)N(C)C1=O |
| IUPAC navn | 5-hex-3-yn-2-yl-1-methyl-5-prop-2-enyl-1,3-diazinane-2,4,6-trione |
| Molekylvægt (g/mol) | 262.3 |
| Formel vægt | 262.1317 |