Få mere at vide
Methocarbamol-d5, TRC
Artikelnummer. 30468677
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30468677 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468677 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2259522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methocarbamol-d5
CAS 1189699-70-4
Renhedsgrad noter HPLC
Molekylær formel C11 2H5 H10 N O5
Mængde 2.5 mg
Synonym [1,1,2,3,3-Pentadeuterio-2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate, 2-Hydroxy-3-(2-methoxyphenoxy)propyl-1,1,2,3,3-d5 carbamate, 3-(2-Methoxyphenoxy)-1-glyceryl-1,1,2,3,3-d5 carbamate, Methocarbamol-d5, Methocarbamol-D5 (glyceryl-D5)
InChI formel InChI=1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14)/i6D2,7D2,8D
SMIL [2H]C([2H])(OC(=O)N)C([2H])(O)C([2H])([2H])Oc1ccccc1OC
IUPAC navn [1,1,2,3,3-pentadeuterio-2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate
Molekylvægt (g/mol) 246.27
Formel vægt 246.1264
Procent renhed >95