Få mere at vide
Methocarbamol-d3, TRC
Artikelnummer. 30480527
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30480527 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480527 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2259535MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methocarbamol-d3
CAS 1346600-86-9
Molekylær formel C11H12D3NO5
Mængde 5 mg
Synonym 3-(2-Methoxyphenoxy)-1,2-propanediol-1-carbamate-d3, 3-(o-Methoxyphenoxy) -2-hydroxypropyl Carbamate-d3, AHR 85-d3, Avetil-d3, Delaxin-d3, Etroflex-d3, Lumirelax-d3, Miolaxene-d3, Neuraxin-d3, Relestrid-d3, Robinax-d3
InChI formel InChI=1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14)/i1D3
SMIL [2H]C([2H])([2H])Oc1ccccc1OCC(O)COC(=O)N
IUPAC navn [2-hydroxy-3-[2-(trideuteriomethoxy)phenoxy]propyl] carbamate
Molekylvægt (g/mol) 244.26
Formel vægt 244.1139