Få mere at vide
Methiocarb-d3 (N-methyl-d3), CDN
Artikelnummer. 30326230
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326230 0.005 g
30317207 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326230 Leverandør CDN Leverandørnr. CDND79660.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methiocarb D3 (N-methyl D3)
CAS 1581694-94-1
Molekylær formel C11 2H3 H12 N O2 S
Mængde 0.005 g
Synonym Methiocarb D3 (N-methyl D3)
InChI formel InChI=1 S/C11H15NO2S/c1-7-5-9(14-11(13)12-3)6-8(2)10(7)15-4/h5-6 H,1-4H3,(H,12,13)/i3D3
SMIL [2 H]C([2 H])([2 H])NC(=O)Oc1cc(C)c(SC)c(C)c1
IUPAC navn (3,5-dimethyl-4-methylsulfanylphenyl) N-(trideuteriomethyl)carbamate
Molekylvægt (g/mol) 228.33
Formel vægt 228.1012 g/mol
Procent renhed 99 atom % D, min 96% Chemical Purity