Få mere at vide
Methimazole-d3, TRC
Artikelnummer. 30473102
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30473102 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473102 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2603021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methimazole-d3
CAS 1160932-07-9
Renhedsgrad noter HPLC
Molekylær formel C4 D3 H3 N2 S
Mængde 1 mg
Synonym 2H-Imidazole-2-thione, 1,3-dihydro-1-(methyl-d3)- (ACI), 1,3-Dihydro-1-(methyl-d3)-2H-imidazole-2-thione (ACI), Methimazole-d3, Methimazole-d3 (methyl-d3), Thiamazole-d3, Thiamazole-D3 (N-methyl-D3)
InChI formel InChI=1S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3H,1H3,(H,5,7)/i1D3
SMIL [2H]C([2H])([2H])N1C=CNC1=S
IUPAC navn 3-(trideuteriomethyl)-1H-imidazole-2-thione
Molekylvægt (g/mol) 117.187
Formel vægt 117.044
Procent renhed >95