Få mere at vide
Methimazole-d3 (methyl-d3), CDN
Artikelnummer. 30318158
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318158 0.01 g
30312095 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318158 Leverandør CDN Leverandørnr. CDND70080.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methimazole-d3
CAS 1160932-07-9
Molekylær formel C4 D3 H3 N2 S
Mængde 0.01 g
Synonym 2 H-Imidazole-2-thione, 1,3-dihydro-1-(methyl-d3)- (ACI), 1,3-Dihydro-1-(methyl-d3)-2 H-imidazole-2-thione (ACI), Methimazole-d3, Methimazole-d3 (methyl-d3), Thiamazole-d3, Thiamazole-D3 (N-methyl-D3)
InChI formel InChI=1 S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3 H,1H3,(H,5,7)/i1D3
SMIL [2 H]C([2 H])([2 H])N1C=CNC1=S
IUPAC navn 3-(trideuteriomethyl)-1 H-imidazole-2-thione
Molekylvægt (g/mol) 117.187
Formel vægt 117.044 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity