Få mere at vide
Methidathion, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methidathion |
| CAS | 950-37-8 |
| Molekylær formel | C6 H11 N2 O4 P S3 |
| Mængde | 1 g |
| Synonym | Phosphorodithioic acid, S-[(5-methoxy-2-oxo-1,3,4-thiadiazol-3(2H)-yl)methyl] O,O-dimethyl ester, Phosphorodithioic acid, O,O-dimethyl ester, S-ester with 4-(mercaptomethyl)-2-methoxy-Δ2-1,3,4-thiadiazolin-5-one (7CI,8CI), DMTP, DMTP (insecticide), Dithiophosphoric acid O,O'-dimethyl-S-[(2-methoxy-1,3,4-thiadiazol-5(4H)-on-4-yl)methyl] ester, Dithiophosphoric acid O,O'-dimethyl-S-[(5-methoxy-1,3,4-thiadiazol-2(3H)-on-3-yl)methyl] ester, GS 13005, Geigy 13005, Geigy GS 13005, Methidathion, Methidation, O,O-Dimethyl S-(5-methoxy-1,3,4-thiadiazole-2 (3H)-on-3-ylmethyl) dithiophosphate, O,O-Dimethyl S-[2-methoxy-1,3,4-thiadiazol-5(4H)-on-4-ylmethyl] phosphorodithioate, O,O-Dimethyl phosphorodithioate S-ester with 4-(mercaptomethyl)-2-methoxy-1,3,4-thiadizaolin-5-one, O,O-Dimethyl-S-(2-methoxy-1,3,4-thiadiazol-5(4H)-onyl-4-methyl) dithiophosphate, O,O-Dimethyl-S-(2-methoxy-Δ2-1,3,4-thiadiazolin-5-on-4-ylmethyl)dithiophosphate, O,O-Dimethyl-S-(5-methoxy-1,3,4-thiadiazol-2(3H)-on-3-ylmethyl)dithiophosphate, O,O-Dimethyl-S-2-methyloxy-1,3,4-thiodiazol-5-(4H)-onyl-(4)-methyldithiophosphate, O,O-Dimethyl-S-[2-methoxy-1,3,4-thiadiazol-5(4H)-onylmethyl] dithiophosphate, O,O-Dimethylphosphorodithioate S-ester with 4-(mercaptomethyl)-2-methoxy-Δ2-1,3,4-thiadiazolin-5-one, O,O'-Dimethyl-S-[(2-methoxy-1,3,4-thiadiazole-5(4H)-one-4-yl)methyl]dithiophosphate, Phosphorodithioic acid O,O'-dimethyl ester S-ester with 4-(mercaptomethyl)-2-methoxy-Δ2-1,3,4-thiadiazolin-5-one, S-((2-Methoxy-5-oxo-Δ2-1,3,4-thiadiazolin-4-yl)methyl) O,O-dimethylphosphorodithioate, Shapulin, Supracid, Supracide, Suprathion, Ultracid, Ultracid 40, Ultracid EC 40 |
| InChI formel | InChI=1S/C6H11N2O4PS3/c1-10-5-7-8(6(9)16-5)4-15-13(14,11-2)12-3/h4H2,1-3H3 |
| SMIL | COC1=NN(CSP(=S)(OC)OC)C(=O)S1 |
| IUPAC navn | 3-(dimethoxyphosphinothioylsulfanylmethyl)-5-methoxy-1,3,4-thiadiazol-2-one |
| Molekylvægt (g/mol) | 302.33 |
| Formel vægt | 301.9619 |