missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metharbital, Mikromol™
Description
Metharbital
Specifications
Specifications
| Kemisk navn eller materiale | Metharbital |
| CAS | 50-11-3 |
| Analyt- eller komponentnavne | Metharbital |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H14 N2 O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 2,4,6(1H,3H,5H)-Pyrimidinetrione, 5,5-diethyl-1-methyl-, Barbituric acid, 5,5-diethyl-1-methyl- (8CI), Metharbital (6CI), 1-Methylbarbital, 5,5-Diethyl-1-methyl-2,4,6(1H,3H,5H)-pyrimidinetrione, 5,5-Diethyl-1-methylbarbituric acid, Endiemal, Endiemalum, Gemonil, Gemonit, Metabarbital, Methabarbitone, Metharbitone, Methylbarbital, N-Methylbarbital, NSC 27156, Sch 412 |
| InChI formel | InChI=1S/C9H14N2O3/c1-4-9(5-2)6(12)10-8(14)11(3)7(9)13/h4-5H2,1-3H3,(H,10,12,14) |
| SMIL | CCC1(CC)C(=O)NC(=O)N(C)C1=O |
| IUPAC navn | 5,5-diethyl-1-methyl-1,3-diazinane-2,4,6-trione |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?