missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Honeywell™

Catalog Number 16615863
Click to view available options
Quantity:
5 L
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Semiconductor grade VLSI (Honeywell 17744)

  • comprehensive portfolio for the electronics and semiconductor industry
  • for scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)
TRUSTED_SUSTAINABILITY
CAS 67-56-1
Molecular Formula CH4O
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Percent Purity min. 99.9 %
Grade Semiconductor
Quantity 5 L
Flash Point 11 °C
Packaging Plastic Bottle
Physical Form Liquid
Water Max. 500 ppm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'