missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99.5%, Pharmpur™ Grade, Scharlab

Catalog Number 11358434
Click to view available options
Quantity:
25 L
Packaging:
HDPE Carboy
This item is not returnable. View return policy
This item is not returnable. View return policy

Extra pure solvent meets Ph Eur, BP and NF specifications, and is analyzed according to guideline CPMP/ICH/283/95.

Scharlab offers the pharmaceutical industry solvents and reagents that comply with the Pharmacopeial Monographies. All Pharmpur™ products comply with Ph Eur, USP and/or other pharmacopeias, indicating that their specifications meet the European or American pharmacopeia.

Melting Point -98°C
Absorbance 0.15 AU at 230nm, 0.05AU at 250nm, 0.02 AU at 270nm, 0.01 AU at 290nm
CAS 67-56-1
Color Colorless
Appearance Clear
Density 0.79g/cm3
Boiling Point 64.5°C
Quantity 25 L
Flash Point 11°C
Identification Passes test
Ignition Point 455°C
Packaging HDPE Carboy
Physical Form Liquid
Refractive Index 1.3288
Residue after Evaporation Max. 0.001%
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
Vapor Pressure 128 hPa
UN Number 1230
Water Max. 0.1%
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Solubility Information Miscible in water
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity Min. 99.5%
Grade Pharmpur™
Chemical Name or Material Methanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'