Learn More
Methanesulfonic acid, silver salt, 99%, Thermo Scientific Chemicals
Catalog Number p-156413
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2386-52-9
CH3AgO3S
202.97
MFCD00064795
MLKQJVFHEUORBO-UHFFFAOYSA-M
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
159658
silver;methanesulfonate
CS(=O)(=O)[O-].[Ag+]
2386-52-9
CH3AgO3S
202.97
MFCD00064795
MLKQJVFHEUORBO-UHFFFAOYSA-M
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
159658
silver;methanesulfonate
CS(=O)(=O)[O-].[Ag+]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 252.0°C to 256.0°C
Color Beige to White or Gray
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula CH3SO3Ag
Beilstein 04,4
Solubility Information Solubility in water: soluble.
Formula Weight 202.97
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Methanesulfonic acid, silver salt, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methanesulfonic acid, silver salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.