Få mere at vide
Methandienone, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methandienone |
| CAS | 72-63-9 |
| Analyt- eller komponentnavne | Methandienone |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H28 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Androsta-1,4-dien-3-one, 17-hydroxy-17-methyl-, (17β)-, Androsta-1,4-dien-3-one, 17β-hydroxy-17-methyl- (8CI), (17β)-17-Hydroxy-17-methylandrosta-1,4-dien-3-one, 1,2-Dehydro-17-methyltestosterone, 1,4-Androstadien-17α-methyl-17β-ol-3-one, 1-Dehydro-17α-methyltestosterone, 1-Dehydromethyltestosterone, 17-Methylandrosta-1,4-dien-17β-ol-3-one, 17α-Methyl-1-dehydrotestosterone, 17α-Methyl-17β-hydroxyandrost-1,4-dien-3-one, 17α-Methyl-17β-hydroxyandrosta-1,4-dien-3-one, 17α-Methylandrosta-1,4-dien-17β-ol-3-one, 17β-Hydroxy-17-methylandrosta-1,4-dien-3-one, 17β-Hydroxy-17α-methyl-1,4-androstadiene-3-one, 17β-Hydroxy-17α-methylandrosta-1,4-dien-3-one, Abirol, Anabolin, Ciba 17309-Ba, Compound 17309, Crein, Danabol, Dehydromethyltestosterone, Dianabol, Dianabole, Geabol, MA, Metanabol, Metandienon, Metandienone, Metandienonum, Metandrostenolon, Metandrostenolone, Metastenol, Methandienone, Methandon, Methandrolone, Methandrostenolone, Methylboldenone, NSC 51180, NSC-42722, Nabolin, Naposim, Nerobol, Nerobolettes, Protobolin, Stenolon, Stenolone, Sterolon, Δ1-17α-Methyltestosterone |
| InChI formel | InChI=1S/C20H28O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,12,15-17,22H,4-5,7-8,10-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1 |
| Løsningstype | Neat |
| SMIL | C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C |
| Vis mere |