Få mere at vide
Metformin EP Impurity C, Mikromol™
Artikelnummer. 17901476
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17901476 25 mg
17914436 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17901476 Leverandør Mikromol Leverandørnr. MM0056.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Metformin EP Impurity C
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine
CAS 1985-46-2
Analyt- eller komponentnavne N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine
Forsendelsestilstand Room Temperature
Molekylær formel C5 H10 N6
Certificeringer/compliance ISO 17025
Synonym N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine, Metformin Hydrochloride Imp. C (EP), N,N-Dimethylmelamine, Metformin Imp. C (EP), Metformin USP Related Compound C, Metformin USP RC C, Metformin Related Compound C, Metformin Hydrochloride Impurity C, Metformin Impurity C
InChI formel InChI=1S/C5H10N6/c1-11(2)5-9-3(6)8-4(7)10-5/h1-2H3,(H4,6,7,8,9,10)
SMIL CN(C)c1nc(N)nc(N)n1
IUPAC navn 2-N,2-N-dimethyl-1,3,5-triazine-2,4,6-triamine
Molekylvægt (g/mol) 154.17
Mængde 25 mg
Formel vægt 154.0967 g/mol