missing translation for 'onlineSavingsMsg'
Få mere at vide

Metformin-d6 Hydrochloride, TRC

Artikelnummer. 30466634
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466634 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466634 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2588125MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Metformin-D6 HCl
CAS 1185166-01-1
Renhedsgrad noter HPLC
Molekylær formel C4 2H6 H5 N5 . Cl H
Mængde 5 mg
Synonym Metformin D6 (dimethyl D6) hydrochloride, Imidodicarbonimidic diamide, N,N-di(methyl-d3)-, hydrochloride (1:1) (ACI), Metformin-d6 monohydrochloride, Metformin D6 (dimethyl D6) hydrochloride, Imidodicarbonimidic diamide, N,N-di(methyl-d3)-, hydrochloride (1:1) (ACI), Metformin-d6 monohydrochloride
InChI formel InChI=1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H/i1D3,2D3;
SMIL Cl.[2H]C([2H])([2H])N(C(=N)NC(=N)N)C([2H])([2H])[2H]
IUPAC navn 3-carbamimidoyl-1,1-bis(trideuteriomethyl)guanidine;hydrochloride
Molekylvægt (g/mol) 171.66
Formel vægt 171.1158
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.