Få mere at vide
Metazosulfuron, TRC
Artikelnummer. 30484265
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30484265 10 mg
30467915 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30484265 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22600510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 868680-84-6
Molekylær formel C15 H18 Cl N7 O7 S
Molekylvægt (g/mol) 475.86
Synonym 1H-Pyrazole-5-sulfonamide, 3-chloro-4-(5,6-dihydro-5-methyl-1,4,2-dioxazin-3-yl)-N-[[(4,6-dimethoxy-2-pyrimidinyl)amino]carbonyl]-1-methyl-, 3-Chloro-N-[(4,6-dimethoxypyrimidin-2-yl)carbamoyl]-1-methyl-4-(5-methyl-5,6-dihydro-1,4,2-dioxazin-3-yl)-1H-pyrazole-5-sulfonamide, Metazosulfuron, NC 620
IUPAC navn 1-[5-chloro-2-methyl-4-(5-methyl-5,6-dihydro-1,4,2-dioxazin-3-yl)pyrazol-3-yl]sulfonyl-3-(4,6-dimethoxypyrimidin-2-yl)urea
SMIL COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2c(C3=NOCC(C)O3)c(Cl)nn2C)n1
Tekniske data
Kemisk navn eller materiale Metazosulfuron
Mængde 10 mg
InChI formel InChI=1S/C15H18ClN7O7S/c1-7-6-29-21-12(30-7)10-11(16)20-23(2)13(10)31(25,26)22-15(24)19-14-17-8(27-3)5-9(18-14)28-4/h5,7H,6H2,1-4H3,(H2,17,18,19,22,24)
Formel vægt 475.0677