Få mere at vide
Metaxalone-d6, TRC
Artikelnummer. 30474185
Skift visning
Mængde:
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30474185 2.5 mg 2.5mg
30477421 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474185 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2258522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metaxalone dimethyl-d6
CAS 1189944-95-3
Molekylær formel C12 2H6 H9 N O3
Mængde 2.5 mg
Synonym Metaxalone-D6, Metaxalone-(dimethyl-D6)
InChI formel InChI=1S/C12H15NO3/c1-8-3-9(2)5-10(4-8)15-7-11-6-13-12(14)16-11/h3-5,11H,6-7H2,1-2H3,(H,13,14)/i1D3,2D3
SMIL [2H]C([2H])([2H])c1cc(OCC2CNC(=O)O2)cc(c1)C([2H])([2H])[2H]
IUPAC navn 5-[[3,5-bis(trideuteriomethyl)phenoxy]methyl]-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 227.29
Formel vægt 227.1429