Få mere at vide
Metaproterenol-d7 Hemisulfate Salt, TRC
Artikelnummer. 30468534
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468534 1 mg
30471503 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468534 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2258371MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346601-49-7
Molekylær formel 2(C11H10D7NO3) • H2O4S
Molekylvægt (g/mol) 2218.309808
Synonym 5-[1-Hydroxy-2-[(1-methylethyl-d7)amino]ethyl]-1,3-benzenediol Sulfate, 3,5-Dihydroxy-α-[(isopropylamino-d7)methyl]benzyl Alcohol Sulfate, (±)-Orciprenaline-d7 Sulfate, 1-(3,5-Dihydroxyphenyl)-1-hydroxy-2-( isopropyl-d7)aminoethane Sulfate, 3,5-Dihydroxy-α-[(isopropylamino- d7)methyl]benzyl Alcohol Sulfate, Alotec-d7, Alupent-d7, Astmopent-d7, Metaprel-d7, Metaproterenol-d7 Sulfate, Novasmasol-d7, Orciprenaline-d7 Sulfate, Orciprenaline-d7 Sulfate, Th 152-d7, dl-Metaproterenol-d7 Sulfate, dl-Orciprenaline-d7 Sulfate
SMIL OC1=CC(C(O)CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=CC(O)=C1.O=S(O)(O)=O
Tekniske data
Kemisk navn eller materiale Metaproterenol-d7 Hemisulfate Salt
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C11H17NO3.H2O4S/c1-7(2)12-6-11(15)8-3-9(13)5-10(14)4-8;1-5(2,3)4/h3-5,7,11-15H,6H2,1-2H3;(H2,1,2,3,4)/i1D3,2D3,7D;
Procent renhed >95%