Få mere at vide
Metaphosphoric Acid Potassium Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Metaphosphoric Acid Potassium Salt |
| CAS | 7790-53-6 |
| Molekylær formel | HKO3P |
| Mængde | 5 g |
| Synonym | Potassium Metaphosphate, Kurrol's Salt, Potassium Kurrol's Salt, Potassium Metaphosphate, Potassium Phosphate (KPO3), Potassium Polymetaphosphate, Phosphenic Acid Potassium Salt |
| InChI formel | InChI=1S/K.HO3P/c;1-4(2)3/h;(H,1,2,3)/q+1;/p-1 |
| SMIL | O=P(O[K])=O |
| Molekylvægt (g/mol) | 118.07 |
| Formel vægt | 117.9222 |