Få mere at vide
Metanicotine, TRC
Artikelnummer. 30465652
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465652 100 mg
30467691 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30465652 Leverandør Toronto Research Chemicals Leverandørnr. TRCM321250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15585-43-0
Molekylær formel C10 H14 N2
Molekylvægt (g/mol) 162.23
Synonym 3-Buten-1-amine, N-methyl-4-(3-pyridinyl)-, (3E)-, 3-Buten-1-amine, N-methyl-4-(3-pyridinyl)-, (E)-, Pyridine, 3-[4-(methylamino)-1-butenyl]-, (E)- (8CI), (3E)-N-Methyl-4-(3-pyridinyl)-3-buten-1-amine, (E)-Metanicotine, RJR 2403, Rivanicline, trans-Metanicotine
IUPAC navn (E)-N-methyl-4-pyridin-3-ylbut-3-en-1-amine
SMIL CNCC\C=C\c1cccnc1
Tekniske data
Kemisk navn eller materiale Metanicotine
Mængde 100 mg
InChI formel InChI=1S/C10H14N2/c1-11-7-3-2-5-10-6-4-8-12-9-10/h2,4-6,8-9,11H,3,7H2,1H3/b5-2+
Formel vægt 162.1157