Få mere at vide
Metanicotine-d3, TRC
Artikelnummer. 30471794
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471794 10 mg
30471005 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471794 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32125210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 220088-40-4
Molekylær formel C10 2H3 H11 N2
Molekylvægt (g/mol) 165.25
Synonym 3-Buten-1-amine, N-(methyl-d3)-4-(3-pyridinyl)-, (3E)- (9CI), Metanicotine-d3, Methyl-d3 Metanicotine, (E)-Metanicotine-D3 (methyl-D3), Rivanicline-D3 (N-methyl-D3), (3E)-N-(Methyl-d3)-4-(3-pyridinyl)-3-buten-1-amine, trans-Metanicotine-d3
IUPAC navn (E)-4-pyridin-3-yl-N-(trideuteriomethyl)but-3-en-1-amine
SMIL [2H]C([2H])([2H])NCC\C=C\c1cccnc1
Tekniske data
Kemisk navn eller materiale (Methyl-d3)metanicotine
Mængde 10 mg
InChI formel InChI=1S/C10H14N2/c1-11-7-3-2-5-10-6-4-8-12-9-10/h2,4-6,8-9,11H,3,7H2,1H3/b5-2+/i1D3
Formel vægt 165.1345