Få mere at vide
Metamizole EP Impurity C (as Hydrochloride), Mikromol™
Beskrivelse
Metamizole EP Impurity C (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Methylaminophenazone Hydrochloride |
| CAS | 856307-27-2 |
| Analyt- eller komponentnavne | 4-Methylaminophenazone Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H15 N3 O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 4-Methylamino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one Hydrochloride (4-Methylaminophenazone Hydrochloride), Metamizole Sodium Monohydrate Imp. C (EP), Metamizole Imp. C (EP), 4-Methylamino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one Hydrochloride, Metamizole Sodium Imp. C (EP) as Hydrochloride, 4-Methylaminophenazone Hydrochloride, Metamizole Sodium Monohydrate Impurity C as Hydrochloride, Metamizole Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C12H15N3O.ClH/c1-9-11(13-2)12(16)15(14(9)3)10-7-5-4-6-8-10;/h4-8,13H,1-3H3;1H |
| SMIL | Cl.CNC1=C(C)N(C)N(C1=O)c2ccccc2 |
| IUPAC navn | 1,5-dimethyl-4-(methylamino)-2-phenylpyrazol-3-one;hydrochloride |
| Vis mere |