Få mere at vide
Metalaxyl-M (~90%), TRC
Artikelnummer. 30006509
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006509 500 mg
17952945 250 mg
17904565 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006509 Leverandør Toronto Research Chemicals Leverandørnr. TRCM258863500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70630-17-0
Molekylær formel C15 H21 N O4
Molekylvægt (g/mol) 279.33
Synonym D-Alanine, N-(2,6-dimethylphenyl)-N-(methoxyacetyl)-, methyl ester (9CI), (-)-(R)-Metalaxyl, (-)-Metalaxyl, (R)-Metalaxyl, Apron XL, CGA 329351, D-(-)-Metalaxyl, Fonganil Gold, Mefenoxam, Metalaxyl-M, R-(-)-Metalaxyl, Ridomil Gold, Ridomil Gold Bravo, Santhal TC, Subdue Maxx, Tega Bion
IUPAC navn methyl (2R)-2-(N-(2-methoxyacetyl)-2,6-dimethylanilino)propanoate
SMIL COCC(=O)N([C@H](C)C(=O)OC)c1c(C)cccc1C
Tekniske data
Kemisk navn eller materiale Metalaxyl-M
Renhedsgrad noter HPLC
Mængde 500 mg
InChI formel InChI=1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3/t12-/m1/s1
Formel vægt 279.1471
Procent renhed >95%