Learn More
VACUUBRAND™ Metal Vacuum Tubing With Small Flanges KF
A durable connection option for the laboratory that still provides flexibility.
Brand: VACUUBRAND™ 20673335
Additional Details : Weight : 25.00000kg
Description
- Manufactured from stainless steel
- Available in a range of sizes and lengths
Specifications
Vacuum Tubing | |
10 mm | |
100 cm | |
10 |
Single bending radius of 19 | |
Metal | |
1 Each |