missing translation for 'onlineSavingsMsg'
Få mere at vide

Quintozene i ethylacetat 1000 μg/ml, Fisher Chemical™

Artikelnummer. 12966023 Shop alle Fisher Chemical produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mL
Pakningsstørrelse:
1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
12966023 1 mL 1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 12966023 Leverandør Fisher Chemical Leverandørnr. F113611

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Alle vores certificerede referencematerialer og reagenser er produceret under en ISO 9001 Quality Management System, også akkrediteret i henhold til ISO/IEC 17025 og akkrediteret iht. ISO Vejledning 34.

Tekniske data

Kemisk navn eller materiale Quintozene (Pentachloronitrobenzene) 
Emballage Ampul
CAS 82-68-8
Molekylær formel C6Cl5NO2
Koncentration 100μ g/ml
FN nummer 1230
InChI nøgle LKPLKUMXSAEKID-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMIL C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-]
IUPAC navn 1,2,3,4,5-pentachlor-6-nitrobenzen
Opløsningsmiddel eller Matrix Methanol
Molekylvægt (g/mol) 295.321
PubChem CID 6720
Mængde 1 mL
ChEBI CHEBI:34908
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.