Learn More
Picloram in Acetonitrile 100μg/mL, Fisher Chemical™
Product Code. 12926413 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
12926413 1 mL 1mL
1 options
Product Code. 12926413 Supplier Fisher Chemical Supplier No. P848500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule
CAS 2-1-1918
Physical Form Liquid
Molecular Formula C6H3Cl3N2O2
Concentration 100 μg/mL
UN Number 1648
InChI Key NQQVFXUMIDALNH-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
IUPAC Name 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 241.45
PubChem CID 15965
Quantity 1 mL
ChEBI CHEBI:34922
Chemical Name or Material Picloram
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Picloram 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .