Learn More
Metabolite Endosulfan-total (sulfate) in Methanol 10μg/mL, Fisher Chemical™
Product Code. 12926633 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. Quantity Unit Size
12926633 10 mL 10mL
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 12926633 Supplier Fisher Chemical Supplier No. P823440
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Specifications
Packaging Ampoule(s)
CAS 1031-07-8
Molecular Formula C9H6Cl6O4S
Concentration 10 μg/mL
InChI Key AAPVQEMYVNZIOO-WINLOITPSA-N
SMILES C1C2C(COS(=O)(=O)O1)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl
Solvent or Matrix Methanol
Molecular Weight (g/mol) 422.903
PubChem CID 71312398
Quantity 10 mL
Chemical Name or Material Endosulfan-total (sulfate)