Learn More
2,4,6-Tribromophenol in DCM 100μg/mL, Fisher Chemical™
Product Code. 12930254 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
12930254 1 mL 1mL
1 options
Product Code. 12930254 Supplier Fisher Chemical Supplier No. P855260
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule
CAS 118-79-6
Physical Form Liquid
Molecular Formula C6H3Br3O
Concentration 100 μg/mL
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES OC1=C(Br)C=C(Br)C=C1Br
IUPAC Name 2,4,6-tribromophenol
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 330.80
PubChem CID 1483
Quantity 1 mL
ChEBI CHEBI:47696
Chemical Name or Material 2,4,6-Tribromophenol