Få mere at vide
meta-Topolin Riboside, TRC
Artikelnummer. 30012023
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012023 100 mg
30012530 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30012023 Leverandør Toronto Research Chemicals Leverandørnr. TRCT540265100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110505-76-5
Molekylær formel C17H19N5O5
Molekylvægt (g/mol) 373.36
Synonym N-[(3-Hydroxyphenyl)methyl]adenosine, Meta-topolin riboside, meta-Topolin-9-riboside, N-(3-Hydroxybenzyl)adenosine, N6-(3-Hydroxybenzyl)adenosine, N6-(m-Hydroxybenzyl)adenosine, N6-(m-Hydroxybenzyl)adenosine, 6-(3-Hydroxybenzylamino)-9-ß-D-ribofuranosylpurine
IUPAC navn (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[(3-hydroxyphenyl)methylamino]purin-9-yl]oxolane-3,4-diol
SMIL OC[C@H]1O[C@H]([C@@H](O)[C@H]1O)n2cnc3c(NCc4cccc(O)c4)ncnc23
Tekniske data
Kemisk navn eller materiale meta-Topolin Riboside
Mængde 100 mg
InChI formel InChI=1S/C17H19N5O5/c23-6-11-13(25)14(26)17(27-11)22-8-21-12-15(19-7-20-16(12)22)18-5-9-2-1-3-10(24)4-9/h1-4,7-8,11,13-14,17,23-26H,5-6H2,(H,18,19,20)/t11-,13+,14+,17-/m1/s1
Formel vægt 373.1386