missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meta-Phosphoric Acid, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
6 x 500 g
Specifications
Concentration | 33.5 to 36.5% |
Quantity | 5 g |
Linear Formula | (HPO3)n |
UN Number | 3260 |
Grade | ACS Reagent |
Chemical Name or Material | Meta-Phosphoric Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'