Få mere at vide
Tocris Bioscience™ Mesulergine hydrochloride
GREENER_CHOICE
Artikelnummer. 18043922
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18043922 10 mg
18035002 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18043922 Leverandør Tocris Bioscience™ Leverandørnr. 1644/10
5-HT2A and 5-HT2C antagonist; also dopamine receptor partial agonist
5-HT2A and 2C receptor antagonist (pA2 values are 9.1 for each receptor) and D2-like dopamine receptor partial agonist (Ki = 8 nM). Displays antiprolactin and antiparkinsonian effects in vivo.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72786-12-0
Molekylær formel C18H27ClN4O2S
Tekniske data
Kemisk navn eller materiale Mesulergine hydrochloride
Mængde 10 mg
Formel vægt 398.95
Renhed >99%
Molekylvægt (g/mol) 398.95