Learn More
Mesothelin Antibody (rMSLN/8764), PE, R&D Systems™
Product Code. 18609114 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity Unit Size
18609114 0.1 mL 0.1mL
1 options
Product Code. 18609114 Supplier Novus Biologicals Supplier No. NBP320892PE
 Restricted Item
Forventet forsendelse: 02-12-2026
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Antigen Mesothelin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMSLN/8764
Conjugate PE
Formulation PBS
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
Host Species Mouse
Immunogen Recombinant fragment (around aa 273-407) of human Mesothelin protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10232
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Product Line Novus