missing translation for 'onlineSavingsMsg'
Få mere at vide

meso-Tetrakis(2-pyridyl)porphine, TRC

Product Code. 30420567
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30420567 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420567 Supplier Toronto Research Chemicals Supplier No. TRCT29854010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale meso-Tetrakis(2-pyridyl)porphine
CAS 40904-90-3
Molekylær formel C40 H26 N8
Mængde 10 mg
Synonym 5,10,15,20-Tetra-2-pyridinyl-21H,23H-porphine, 5,10,15,20-Tetra-2-pyridylporphine, 5,10,15,20-Tetrakis(2-pyridyl)porphyrin, meso-Tetra-2-pyridylporphine, meso-Tetrakis(2-pyridyl)porphyrin, meso-Tetrakis(o-pyridyl)porphine
InChI formel InChI=1S/C40H26N8/c1-5-21-41-25(9-1)37-29-13-15-31(45-29)38(26-10-2-6-22-42-26)33-17-19-35(47-33)40(28-12-4-8-24-44-28)36-20-18-34(48-36)39(27-11-3-7-23-43-27)32-16-14-30(37)46-32/h1-24,45,48H/b37-29-,37-30-,38-31-,38-33-,39-32-,39-34-,40-35-,40-36-
SMIL C1=Cc2[nH]c1c(c3ccccn3)c4ccc(n4)c(c5ccccn5)c6ccc([nH]6)c(c7ccccn7)c8ccc(n8)c2c9ccccn9
IUPAC navn (5E,10Z,14Z,19E)-5,10,15,20-tetrapyridin-2-yl-21,23-dihydroporphyrin
Molekylvægt (g/mol) 618.688
Formel vægt 618.228
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.