missing translation for 'onlineSavingsMsg'
Få mere at vide

Mesaconsyre, TRC

Artikelnummer. 30474953
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474953 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30474953 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2256901G
 Begrænset vare
Forventet forsendelse: 10-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Citronic acid
CAS 498-24-8
Molekylær formel C5 H6 O4
Mængde 1 g
Synonym 2-Butenedioic acid, 2-methyl-, (2E)-, 2-Butenedioic acid, 2-methyl-, (E)-, Mesaconic acid (6CI,8CI), (2E)-2-Methyl-2-butenedioic acid, (E)-Citraconic acid, 2-Methylfumaric acid, Methylfumaric acid, NSC 65438, trans-1-Propene-1,2-dicarboxylic acid, trans-2-Methyl-2-butenedioic acid
InChI formel InChI=1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h2H,1H3,(H,6,7)(H,8,9)/b3-2+
SMIL C\C(=C/C(=O)O)\C(=O)O
IUPAC navn (E)-2-methylbut-2-enedioinsyre
Molekylvægt (g/mol) 130.0987
Formel vægt 130.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.