Få mere at vide
Meropenem-d6 (>90%), TRC
Artikelnummer. 30470721
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30470721 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470721 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22561710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Meropenem-d6
CAS 1217976-95-8
Renhedsgrad noter HPLC
Molekylær formel C17H19D6N3O5S
Mængde 10 mg
Synonym (4R,5S,6S)-3-[[3S,5S)-5-[(Dimethylamino-d6)carbonyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic Acid, ICI 194660-d6, Mepem-d6, Meronem-d6, Meropen-d6, Merrem-d6, Merrem IV-d6, SM 7338-d6
InChI formel InChI=1S/C17H25N3O5S/c1-7-12-11(8(2)21)16(23)20(12)13(17(24)25)14(7)26-9-5-10(18-6-9)15(22)19(3)4/h7-12,18,21H,5-6H2,1-4H3,(H,24,25)/t7-,8-,9+,10+,11-,12-/m1/s1/i3D3,4D3
SMIL O=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])[C@H](C1)NC[C@H]1SC2=C(C(O)=O)N3C([C@]([H])([C@H](O)C)[C@@]3([H])[C@H]2C)=O
Molekylvægt (g/mol) 389.5
Procent renhed >95