missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury, Certified AR for Analysis, Metal, Fisher Chemical™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Specifications
Color | Silver |
Melting Point | -38.9°C |
Boiling Point | 356.5°C |
Quantity | 100 g |
Formula Weight | 200.59g/mol |
Packaging | HDPE plastic bottle |
Vapor Pressure | 0.01 hPa at 20°C |
Form | Liquid |
Safety and Handling
Product Identifier
- Mercury
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360D-May damage the unborn child.
- H330-Fatal if inhaled.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'