missing translation for 'onlineSavingsMsg'
Learn More

Mercury, Certified AR for Analysis, Metal, Fisher Chemical™

Brand:  Fisher Chemical M/3750/60

Additional Details : CAS Number : 7439-97-6 Weight : 1.00000kg Transport : UN number : 2809 Chem class : 8 Pack group : III


Quantity 1kg

 View more versions of this product

Product Code. 10438030

  • POA / 1kg

Currently not in stock

Please to purchase this item. Need a web account? Register with us today!

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on +45 70 27 99 20 for assistance.
View alternative products »


Description

Description

Specifications

Chemical Identifiers

7439-97-6
200.592
QSHDDOUJBYECFT-UHFFFAOYSA-N
23931
mercury
Hg
11035
hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
CHEBI:16170
[Hg]

Specifications

1kg
Hg
hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
[Hg]
200.592
23931
200.59g/mol
0.01 hPa at 20°C
Silver
356.5°C
7439-97-6
11035
QSHDDOUJBYECFT-UHFFFAOYSA-N
mercury
HDPE plastic bottle
CHEBI:16170
Liquid
7
-38.9°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Mercury

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1

Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360D-May damage the unborn child.
  • H330-Fatal if inhaled.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents