missing translation for 'onlineSavingsMsg'
Learn More

Mercury, Certified AR for Analysis, Metal, Fisher Chemical™

Catalog Number 10253960
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
This item is not returnable. View return policy
7439-97-6
Hg
200.59
MFCD00011035
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
23931
CHEBI:16170
mercury
[Hg]
This item is not returnable. View return policy
7439-97-6
Hg
200.59
MFCD00011035
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
23931
CHEBI:16170
mercury
[Hg]
Color Silver
Melting Point -38.9°C
Boiling Point 356.5°C
Quantity 500 g
Formula Weight 200.59g/mol
Packaging HDPE plastic bottle
Vapor Pressure 0.01 hPa at 20°C
Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360D-May damage the unborn child.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'