missing translation for 'onlineSavingsMsg'
Learn More
Mercury, AAS standard solution, Specpure™ Hg 1000μg/mL Product Code.: 11389169

Mercury, AAS standard solution, Specpure™ Hg 1000μg/mL

Product Code. p-7080421
100 mL, 100mL
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11389169

Brand: Thermo Scientific Alfa Aesar 088079.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00011035

Spécification

Physical Form Liquid
Concentration Hg in 5% HNO3
UN Number UN3264
Quantity 100 mL
Grade Specpure™
Chemical Name or Material Mercury, AAS standard solution, Hg 1000ÎĽg/mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number1523238 Certificate TypeCertificate of Analysis Date22/01/2025 Product Code088079.AP,088079.AE
Lot Number1523235 Certificate TypeCertificate of Analysis Date22/01/2025 Product Code088079.AP,088079.AE
Lot Number1502147 Certificate TypeCertificate of Analysis Date19/11/2024 Product Code088079.AP,088079.AE
Lot Number1502115 Certificate TypeCertificate of Analysis Date19/11/2024 Product Code088079.AP,088079.AE
Lot Number1504895 Certificate TypeCertificate of Analysis Date19/11/2024 Product Code088079.AP,088079.AE

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Mercury, AAS standard solution, Specpure™ Hg 1000μg/mL >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis