missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid calcium salt trihydrate, 98%, Thermo Scientific™

Catalog Number 15482357
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
65208-41-5
C2H10CaO5S
186.24
MFCD00150707
GDECCIJZCWLFSB-UHFFFAOYSA-N
calcium,2-sulfidoacetate,trihydrate, thioglycolic acid calcium trihydrate
131882808
calcium;2-sulfanylacetic acid;trihydrate
O.O.O.[Ca].OC(=O)CS
This item is not returnable. View return policy
65208-41-5
C2H10CaO5S
186.24
MFCD00150707
GDECCIJZCWLFSB-UHFFFAOYSA-N
calcium,2-sulfidoacetate,trihydrate, thioglycolic acid calcium trihydrate
131882808
calcium;2-sulfanylacetic acid;trihydrate
O.O.O.[Ca].OC(=O)CS

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 280°C to 290°C (decomposition)
Color White
Linear Formula (HSCH2CO2)Ca·3H2O
Quantity 10 g
Beilstein 3755791
Merck Index 14,1711
Solubility Information Soluble in water.
Formula Weight 276.34 (222.29 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mercaptoacetic acid calcium salt trihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'