missing translation for 'onlineSavingsMsg'
Få mere at vide

Meptyldinocap, TRC

Artikelnummer. 30466899
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466899 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30466899 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2251301MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Meptyldinocap
CAS 131-72-6
Molekylær formel C18 H24 N2 O6
Mængde 1 mg
Synonym Dinocap-P, 2,4-Dinitro-6-sec-butylphenyl crotonate
InChI formel InChI=1S/C18H24N2O6/c1-4-6-7-8-10-13(3)15-11-14(19(22)23)12-16(20(24)25)18(15)26-17(21)9-5-2/h5,9,11-13H,4,6-8,10H2,1-3H3/b9-5+
SMIL CCCCCCC(C)c1cc(cc(c1OC(=O)\C=C\C)[N+](=O)[O-])[N+](=O)[O-]
IUPAC navn (2,4-dinitro-6-octan-2-ylphenyl) (E)-but-2-enoate
Molekylvægt (g/mol) 364.39
Formel vægt 364.1634
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.