missing translation for 'onlineSavingsMsg'
Få mere at vide

Meptyldinocap, TRC

Product Code. 30476429
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30476429 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30476429 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM22513010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Meptyldinocap
CAS 131-72-6
Molekylær formel C18 H24 N2 O6
Mængde 10 mg
Synonym Dinocap-P, 2,4-Dinitro-6-sec-butylphenyl crotonate
InChI formel InChI=1S/C18H24N2O6/c1-4-6-7-8-10-13(3)15-11-14(19(22)23)12-16(20(24)25)18(15)26-17(21)9-5-2/h5,9,11-13H,4,6-8,10H2,1-3H3/b9-5+
SMIL CCCCCCC(C)c1cc(cc(c1OC(=O)\C=C\C)[N+](=O)[O-])[N+](=O)[O-]
IUPAC navn (2,4-dinitro-6-octan-2-ylphenyl) (E)-but-2-enoate
Molekylvægt (g/mol) 364.39
Formel vægt 364.1634
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.