Få mere at vide
Meprobamate, Mikromol
Artikelnummer. 30457886
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30457886 50 mg
1 options
Artikelnummer. 30457886 Leverandør Mikromol Leverandørnr. MM0929.00
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Meprobamate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Meprobamate
CAS 57-53-4
Analyt- eller komponentnavne Meprobamate
Forsendelsestilstand Room Temperature
Molekylær formel C9 H18 N2 O4
Certificeringer/compliance ISO 17025
Synonym Meprobamate, 2-Methyl-2-propylpropane-1,3-diyl dicarbamate, 2-Methyl-2-propyl-1,3-propanediol 1,3-dicarbamate, 2-Methyl-2-propyltrimethylene dicarbamate, 2-Methyl-2-propyl-1,3-propanediol dicarbamate, 2,2-Di(carbamoyloxymethyl)pentane, 2-Methyl-2-n-propyl-1,3-propanediol dicarbamate, 2-Methyl-2-propyl-1,3-propanediol dicarbamate, Amepromat, Amosene, Anastress, Anatimon, Andaxin, Aneural, Ansiatan, Anzil, Apascil, Appetrol, Arcoban, Artolon, Atraxin, Ayeramate, Bamo 400, Biobamat, Biobamate, Calmax, Calmiren, Canquil-400, Cap-O-Tran, Carb-A-Med, Carbamic acid 2-methyl-2-propyltrimethylene ester, Carbaxin, Cirpon, Crestanil, Cyrpon, Dicandiol, Diron, Ecuanil, Epikur, Equanil, Equanil Suspension, Equinil, Fas-Cile 200, Gadexyl, Harmonin, Hartol, Holbamate, Kesso-Bamate, Klort, Larten, Lepetown, Libiolan, Mar-Bate, Mepantin, Mepavlon, Meposed, Mepr, Meprin, Meprindon, Meprobam, Meprobamat, Meprobamat-Petrasch, Meprobamate, Meproban, Meprocompren, Meprocon CMC, Meprol, Mepronil, Meprosin, Meprospan, Meprotabs, Meprotan, Meproten, Meprotil, Meptran, Mesmar, Metranquil, Micrainin, Microbamat, Milprem, Miltaun, Miltown, Miltrate, Morbam, My-trans, NSC 30418, Nervonus, Oasil, PMB 200, PMB 400, Panediol, Perequil, Perquietil, Pertranquil, Placidon, Placitate, Probamyl, Procalmadiol, Procalmidol, Promate, Protran, Quaname, Quanil, Rastenil, Reostral, Restenil, Robamate, Sedanil, Sedazil, Seril, Setran, Sowell, Tamate, Trankvilan, Tranlisant, Tranquilan, Tranquilax, Tranquiline, Urbil, Urbilat, Vio-Bamate, Carisoprodol Impurity D
InChI formel InChI=1S/C9H18N2O4/c1-3-4-9(2,5-14-7(10)12)6-15-8(11)13/h3-6H2,1-2H3,(H2,10,12)(H2,11,13)
Løsningstype Neat
SMIL CCCC(C)(COC(=O)N)COC(=O)N
IUPAC navn [2-(carbamoyloxymethyl)-2-methylpentyl] carbamate
Molekylvægt (g/mol) 218.25
Mængde 50 mg
Formel vægt 218.1267 g/mol